6-Bromopyrazolo[1,5-a]pyridine 98%
Dodavatel: Apollo Scientific
OR910028-1G
APOSOR910028-1GEA
3270
CZK
APOSOR910028-1G
6-Bromopyrazolo[1,5-a]pyridine 98%
6-Bromopyrazolo[1,5-a]pyridine
Vzorec:
C₇H₅BrN₂ MW: 197,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18633056 CAS číslo: 1264193-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...