5-Chloro-4-Hydroxy-8-methylquinoline 95%
Dodavatel: Apollo Scientific
OR909850-1G
OR909850-5G
APOSOR909850-1GEA
4140
CZK
APOSOR909850-1G
APOSOR909850-5G
5-Chloro-4-Hydroxy-8-methylquinoline 95%
5-Chloro-4-Hydroxy-8-methylquinoline
Vzorec:
C₁₀H₈ClNO MW: 193.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00272438 CAS číslo: 203626-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...