6-Hydroxy-2-methylquinazolin-4(3H)-one 98%
Dodavatel: Apollo Scientific
OR909804-1G
OR909804-250MG
APOSOR909804-1GEA
10350
CZK
APOSOR909804-1G
APOSOR909804-250MG
6-Hydroxy-2-methylquinazolin-4(3H)-one 98%
6-Hydroxy-2-methylquinazolin-4(3H)-one
Vzorec:
C₉H₈N₂O₂ MW: 176,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01685973 CAS číslo: 1882-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...