Tisk…

2H-1,3-benzodioxol-4-ol 98%

Dodavatel: Apollo Scientific

Synonyms: Benzo[d][1,3]dioxol-4-ol

OR909770-1G
APOSOR909770-1GEA 3060 CZK
APOSOR909770-1G
2H-1,3-benzodioxol-4-ol 98%
2H-1,3-benzodioxol-4-ol
Vzorec: C₇H₆O₃
MW: 138,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092232
CAS číslo: 69393-72-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR