Tisk…

1-(Methoxymethyl)pyrrole 98%

Dodavatel: Apollo Scientific

OR909719-10G OR909719-1G OR909719-5G
APOSOR909719-10GEA 25630 CZK
APOSOR909719-10G APOSOR909719-1G APOSOR909719-5G
1-(Methoxymethyl)pyrrole 98%
1-(Methoxymethyl)pyrrole
Vzorec: C₆H₉NO
MW: 111,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923696
CAS číslo: 76322-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR