6-Chloro-2-methoxypyridin-3-ol 98%
Dodavatel: Apollo Scientific
OR909718-1G
OR909718-250MG
APOSOR909718-1GEA
16340
CZK
APOSOR909718-1G
APOSOR909718-250MG
6-Chloro-2-methoxypyridin-3-ol 98%
6-Chloro-2-methoxypyridin-3-ol
Vzorec:
C₆H₆ClNO₂ MW: 159,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189892 CAS číslo: 1196146-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...