Tisk…

N-Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxamide 98%

Dodavatel: Apollo Scientific

OR909710-10G OR909710-1G OR909710-5G
APOSOR909710-10GEA 21490 CZK
APOSOR909710-10G APOSOR909710-1G APOSOR909710-5G
N-Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxamide 98%
N-Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxamide
Vzorec: C₁₀H₁₁NO₃
MW: 193,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19597438
CAS číslo: 1203954-92-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR