1-Boc-3-acetyl-piperidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl 3-acetylpiperidine-1-carboxylate
OR909709-1G
OR909709-5G
APOSOR909709-1GEA
787
CZK
APOSOR909709-1G
APOSOR909709-5G
1-Boc-3-acetyl-piperidine 97%
1-Boc-3-acetyl-piperidine
Vzorec:
C₁₂H₂₁NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09953372 CAS číslo: 858643-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...