Tisk…
1-(Piperidin-3-yl)ethanone hydrochloride 98%
Dodavatel: Apollo Scientific
OR909708-1G OR909708-5G
APOSOR909708-1GEA 9330 CZK
APOSOR909708-1G APOSOR909708-5G
1-(Piperidin-3-yl)ethanone hydrochloride 98%
1-(Piperidin-3-yl)ethanone hydrochloride
Vzorec: C₇H₁₃NO·ClH
MW: 163,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022655
CAS číslo: 89895-05-6