2-Methyl-5-(2-methylphenyl)pyrazol-3-amine 97%
Dodavatel: Apollo Scientific
OR909664-1G
OR909664-5G
APOSOR909664-1GEA
6220
CZK
APOSOR909664-1G
APOSOR909664-5G
2-Methyl-5-(2-methylphenyl)pyrazol-3-amine 97%
2-Methyl-5-(2-methylphenyl)pyrazol-3-amine
Vzorec:
C₁₁H₁₃N₃ MW: 187,241 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06637387 CAS číslo: 1017665-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...