Tisk…

Oxazolo[4,5-b]pyridine 96%

Dodavatel: Apollo Scientific

OR909642-1G OR909642-5G
APOSOR909642-1GEA 2820 CZK
APOSOR909642-1G APOSOR909642-5G
Oxazolo[4,5-b]pyridine 96%
Oxazolo[4,5-b]pyridine
Vzorec: C₆H₄N₂O
MW: 120,109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13175183
CAS číslo: 273-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR