Tisk…

4-Methyl-2-(methylamino)thiazole 98%

Dodavatel: Apollo Scientific

OR909624-1G OR909624-5G
APOSOR909624-1GEA 8270 CZK
APOSOR909624-1G APOSOR909624-5G
4-Methyl-2-(methylamino)thiazole 98%
4-Methyl-2-(methylamino)thiazole
Vzorec: C₅H₈N₂S
Storage Temperature: Ambient
MDL Number: MFCD01924766
CAS číslo: 2161-68-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR