Tisk…

tert-Butyl N-cyclopropyl-N-methylcarbamate 97%

Dodavatel: Apollo Scientific

OR909590-1G OR909590-5G
APOSOR909590-1GEA 4630 CZK
APOSOR909590-1G APOSOR909590-5G
tert-Butyl N-cyclopropyl-N-methylcarbamate 97%
tert-Butyl N-cyclopropyl-N-methylcarbamate
Vzorec: C₉H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD21105448
CAS číslo: 1378390-62-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR