Tisk…

4-Benzylmorpholine 97%

Dodavatel: Apollo Scientific

OR909577-10G OR909577-1G OR909577-5G
APOSOR909577-10GEA 5630 CZK
APOSOR909577-10G APOSOR909577-1G APOSOR909577-5G
4-Benzylmorpholine 97%
4-Benzylmorpholine
Vzorec: C₁₁H₁₅NO
MW: 177,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00090503
CAS číslo: 10316-00-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR