Methyl 1-propylpyrrole-2-carboxylate 95%
Dodavatel: Apollo Scientific
OR909560-1G
OR909560-25G
OR909560-5G
APOSOR909560-1GEA
4140
CZK
APOSOR909560-1G
APOSOR909560-25G
APOSOR909560-5G
Methyl 1-propylpyrrole-2-carboxylate 95%
Methyl 1-propylpyrrole-2-carboxylate
Vzorec:
C₉H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22421508 CAS číslo: 70696-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...