Tisk…

Methyl 1-ethylpyrrole-2-carboxylate 97%

Dodavatel: Apollo Scientific

OR909559-1G OR909559-5G
APOSOR909559-1GEA 4770 CZK
APOSOR909559-1G APOSOR909559-5G
Methyl 1-ethylpyrrole-2-carboxylate 97%
Methyl 1-ethylpyrrole-2-carboxylate
Vzorec: C₈H₁₁NO₂
MW: 153,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13174842
CAS číslo: 73058-14-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR