Tisk…

N-Butyl-2-chloropyrimidin-4-amine 95%

Dodavatel: Apollo Scientific

OR909540-1G OR909540-5G
APOSOR909540-1GEA 7670 CZK
APOSOR909540-1G APOSOR909540-5G
N-Butyl-2-chloropyrimidin-4-amine 95%
N-Butyl-2-chloropyrimidin-4-amine
Vzorec: C₈H₁₂ClN₃
MW: 185,654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16712456
CAS číslo: 66229-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR