2-Ethyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
Dodavatel: Apollo Scientific
OR909487-10G
OR909487-1G
OR909487-5G
APOSOR909487-10GEA
21490
CZK
APOSOR909487-10G
APOSOR909487-1G
APOSOR909487-5G
2-Ethyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
2-Ethyl-2,4-dihydro-1,4-benzoxazin-3-one
Vzorec:
C₁₀H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03830297 CAS číslo: 90921-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...