1-(3-Chlorophenyl)pyrrolidin-2-one 98%
Dodavatel: Apollo Scientific
OR909413-10G
OR909413-1G
OR909413-5G
APOSOR909413-10GEA
21490
CZK
APOSOR909413-10G
APOSOR909413-1G
APOSOR909413-5G
1-(3-Chlorophenyl)pyrrolidin-2-one 98%
1-(3-Chlorophenyl)pyrrolidin-2-one
Vzorec:
C₁₀H₁₀ClNO MW: 195,646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00522616 CAS číslo: 24051-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...