Tisk…

2-Ethoxy-3-propoxypyridine 98%

Dodavatel: Apollo Scientific

OR909396-1G OR909396-25G OR909396-5G
APOSOR909396-1GEA 3540 CZK
APOSOR909396-1G APOSOR909396-25G APOSOR909396-5G
2-Ethoxy-3-propoxypyridine 98%
2-Ethoxy-3-propoxypyridine
Vzorec: C₁₀H₁₅NO₂
MW: 181,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981543
CAS číslo: 1330750-35-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR