Tisk…

N-Ethyl-1-methylpyrrole-2-carboxamide 97%

Dodavatel: Apollo Scientific

OR909321-1G OR909321-25G OR909321-5G
APOSOR909321-1GEA 3540 CZK
APOSOR909321-1G APOSOR909321-25G APOSOR909321-5G
N-Ethyl-1-methylpyrrole-2-carboxamide 97%
N-Ethyl-1-methylpyrrole-2-carboxamide
Vzorec: C₈H₁₂N₂O
MW: 152,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18087710
CAS číslo: 124725-22-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR