Tisk…

N-Ethyl-1H-pyrrole-2-carboxamide 98%

Dodavatel: Apollo Scientific

OR909320-1G OR909320-25G OR909320-5G
APOSOR909320-1GEA 3110 CZK
APOSOR909320-1G APOSOR909320-25G APOSOR909320-5G
N-Ethyl-1H-pyrrole-2-carboxamide 98%
N-Ethyl-1H-pyrrole-2-carboxamide
Vzorec: C₇H₁₀N₂O
MW: 138,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143556
CAS číslo: 156741-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR