N-Methyl 1-methylpyrrole-2-carboxamide 97%
Dodavatel: Apollo Scientific
OR909319-1G
OR909319-5G
APOSOR909319-1GEA
4770
CZK
APOSOR909319-1G
APOSOR909319-5G
N-Methyl 1-methylpyrrole-2-carboxamide 97%
N-Methyl 1-methylpyrrole-2-carboxamide
Vzorec:
C₇H₁₀N₂O MW: 138,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18087709 CAS číslo: 7151-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...