Tisk…

N-Methyl-1H-pyrrole-2-carboxamide 97%

Dodavatel: Apollo Scientific

OR909318-1G OR909318-5G
APOSOR909318-1GEA 4140 CZK
APOSOR909318-1G APOSOR909318-5G
N-Methyl-1H-pyrrole-2-carboxamide 97%
N-Methyl-1H-pyrrole-2-carboxamide
Vzorec: C₆H₈N₂O
MW: 124,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143555
CAS číslo: 132911-42-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR