Tisk…

2-Methyl-4-nitroanisole 98%

Dodavatel: Apollo Scientific

Synonyms: 1-Methoxy-2-methyl-4-nitrobenzene

OR909315-100G
APOSOR909315-100GEA 2360 CZK
APOSOR909315-100G
2-Methyl-4-nitroanisole 98%
2-Methyl-4-nitroanisole
Vzorec: C₈H₉NO₃
MW: 167.16
Bod varu: 311
Bod tání: 96
Storage Temperature: Ambient
MDL Number: MFCD07787583
CAS číslo: 50741-92-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR