Tisk…

N,4-Dimethyl-2-nitroaniline 95%

Dodavatel: Apollo Scientific

OR909293-1G OR909293-250MG
APOSOR909293-1GEA 16340 CZK
APOSOR909293-1G APOSOR909293-250MG
N,4-Dimethyl-2-nitroaniline 95%
N,4-Dimethyl-2-nitroaniline
Vzorec: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07437778
CAS číslo: 4600-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR