Tisk…

3-Isobutyl-1H-pyrazole 97%

Dodavatel: Apollo Scientific

OR909282-1G OR909282-5G
APOSOR909282-1GEA 5200 CZK
APOSOR909282-1G APOSOR909282-5G
3-Isobutyl-1H-pyrazole 97%
3-Isobutyl-1H-pyrazole
Vzorec: C₇H₁₂N₂
MW: 124,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701141
CAS číslo: 98816-40-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR