Tisk…

4-Bromo-3-cyclopropyl-1H-pyrazole 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-5-cyclopropyl-1H-pyrazole

OR909255-1G OR909255-5G
APOSOR909255-1GEA 3020 CZK
APOSOR909255-1G APOSOR909255-5G
4-Bromo-3-cyclopropyl-1H-pyrazole 95%
4-Bromo-3-cyclopropyl-1H-pyrazole
Vzorec: C₆H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD14658056
CAS číslo: 957345-28-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR