Tisk…

(Z)-3-Cyclopropylbut-2-enoic acid 98%

Dodavatel: Apollo Scientific

OR909195-1G OR909195-5G
APOSOR909195-1GEA 6220 CZK
APOSOR909195-1G APOSOR909195-5G
(Z)-3-Cyclopropylbut-2-enoic acid 98%
(Z)-3-Cyclopropylbut-2-enoic acid
Vzorec: C₇H₁₀O₂
MW: 126,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756435
CAS číslo: 766-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR