Methyl 2-amino-4-cyclopropylpyrimidine-5-carboxylate 95%
Dodavatel: Apollo Scientific
OR909027-10G
OR909027-1G
OR909027-5G
APOSOR909027-10GEA
21490
CZK
APOSOR909027-10G
APOSOR909027-1G
APOSOR909027-5G
Methyl 2-amino-4-cyclopropylpyrimidine-5-carboxylate 95%
Methyl 2-amino-4-cyclopropylpyrimidine-5-carboxylate
Vzorec:
C₉H₁₁N₃O₂ MW: 193,203 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11855863 CAS číslo: 1133115-42-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...