Tisk…

4-(4-Chlorophenyl)-1H-1,2,4-triazol-5(4H)-one 98%

Dodavatel: Apollo Scientific

OR908964-1G OR908964-25G OR908964-5G
APOSOR908964-1GEA 4140 CZK
APOSOR908964-1G APOSOR908964-25G APOSOR908964-5G
4-(4-Chlorophenyl)-1H-1,2,4-triazol-5(4H)-one 98%
4-(4-Chlorophenyl)-1H-1,2,4-triazol-5(4H)-one
Vzorec: C₈H₆ClN₃O
MW: 195,606 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053882
CAS číslo: 5097-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR