Tisk…

4-(2-Pyridyl)phenylboronic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(Pyridin-2-yl)phenylboronic acid, (4-(Pyridin-2-yl)phenyl)boronic acid

OR908412-1G OR908412-5G
APOSOR908412-1GEA 2110 CZK
APOSOR908412-1G APOSOR908412-5G
4-(2-Pyridyl)phenylboronic acid 95%
4-(2-Pyridyl)phenylboronic acid
Vzorec: C₁₁H₁₀BNO₂
MW: 199,014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17013972
CAS číslo: 170230-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR