(6-Cyclopropylpyridin-3-yl)boronic acid ≥98%
Dodavatel: Apollo Scientific
OR908405-100MG
OR908405-1G
OR908405-250MG
APOSOR908405-100MGEA
2690
CZK
APOSOR908405-100MG
APOSOR908405-1G
APOSOR908405-250MG
(6-Cyclopropylpyridin-3-yl)boronic acid ≥98%
(6-Cyclopropylpyridin-3-yl)boronic acid
Vzorec:
C₈H₁₀BNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18255555 CAS číslo: 1253055-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...