Tisk…

2-(Cyclopropylmethoxy)pyridine-3-boronic acid 98%

Dodavatel: Apollo Scientific

OR908399-1G OR908399-5G
APOSOR908399-1GEA 8270 CZK
APOSOR908399-1G APOSOR908399-5G
2-(Cyclopropylmethoxy)pyridine-3-boronic acid 98%
2-(Cyclopropylmethoxy)pyridine-3-boronic acid
Vzorec: C₉H₁₂BNO₃
MW: 193.007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258839
CAS číslo: 1621416-45-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR