Tisk…

tert-Butyl N-[2-(dimethylamino)ethyl]carbamate 96%

Dodavatel: Apollo Scientific
Varování

OR908338-1G OR908338-5G
APOSOR908338-1GEA 6450 CZK
APOSOR908338-1G APOSOR908338-5G
tert-Butyl N-[2-(dimethylamino)ethyl]carbamate 96%
tert-Butyl N-[2-(dimethylamino)ethyl]carbamate
Vzorec: C₉H₂₀N₂O₂
MW: 188,267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08729296
CAS číslo: 196200-04-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR