Benzyl 3-chloropropanoate 96%
Dodavatel: Apollo Scientific
OR908220-1G
OR908220-5G
APOSOR908220-1GEA
4140
CZK
APOSOR908220-1G
APOSOR908220-5G
Benzyl 3-chloropropanoate 96%
Benzyl 3-chloropropanoate
Vzorec:
C₁₀H₁₁ClO₂ MW: 198,646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20542317 CAS číslo: 6282-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...