Tisk…

4-Cyano-N-methylbenzamide 98%

Dodavatel: Apollo Scientific

OR908189-1G OR908189-5G
APOSOR908189-1GEA 6220 CZK
APOSOR908189-1G APOSOR908189-5G
4-Cyano-N-methylbenzamide 98%
4-Cyano-N-methylbenzamide
Vzorec: C₉H₈N₂O
MW: 160,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03550597
CAS číslo: 36268-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR