4-(Isopropylsulphanyl)benzoic acid 96%
Dodavatel: Apollo Scientific
OR908181-1G
OR908181-5G
APOSOR908181-1GEA
7670
CZK
APOSOR908181-1G
APOSOR908181-5G
4-(Isopropylsulphanyl)benzoic acid 96%
4-(Isopropylsulphanyl)benzoic acid
Vzorec:
C₁₀H₁₂O₂S MW: 196,266 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09735126 CAS číslo: 13205-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...