Tisk…

Methyl 3-formyl-5-methylbenzoate 97%

Dodavatel: Apollo Scientific
Varování

OR908164-1G OR908164-250MG
APOSOR908164-1GEA 25530 CZK
APOSOR908164-1G APOSOR908164-250MG
Methyl 3-formyl-5-methylbenzoate 97%
Methyl 3-formyl-5-methylbenzoate
Vzorec: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544349
CAS číslo: 1205514-72-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR