Tisk…

3-(Oxolan-2-ylmethoxy)propanenitrile 96%

Dodavatel: Apollo Scientific

OR908150-10G OR908150-1G OR908150-5G
APOSOR908150-10GEA 28540 CZK
APOSOR908150-10G APOSOR908150-1G APOSOR908150-5G
3-(Oxolan-2-ylmethoxy)propanenitrile 96%
3-(Oxolan-2-ylmethoxy)propanenitrile
Vzorec: C₈H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00466610
CAS číslo: 5338-10-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR