4-Hydroxy-3-methylbenzonitrile 98%
Dodavatel: Apollo Scientific
OR908148-1G
OR908148-5G
APOSOR908148-1GEA
0
CZK
APOSOR908148-1G
APOSOR908148-5G
4-Hydroxy-3-methylbenzonitrile 98%
4-Hydroxy-3-methylbenzonitrile
Vzorec:
C₈H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD11848762 CAS číslo: 15777-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...