2-Hydroxy-3-methyl-2-phenylbutanoic acid 98%
Dodavatel: Apollo Scientific
OR908116-1G
OR908116-250MG
APOSOR908116-1GEA
16340
CZK
APOSOR908116-1G
APOSOR908116-250MG
2-Hydroxy-3-methyl-2-phenylbutanoic acid 98%
2-Hydroxy-3-methyl-2-phenylbutanoic acid
Vzorec:
C₁₁H₁₄O₃ MW: 194,227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00021679 CAS číslo: 15879-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...