Tisk…

2-Bromo-4-hydroxybenzonitrile 98%

Dodavatel: Apollo Scientific

OR908076-1G OR908076-250MG
APOSOR908076-1GEA 7280 CZK
APOSOR908076-1G APOSOR908076-250MG
2-Bromo-4-hydroxybenzonitrile 98%
2-Bromo-4-hydroxybenzonitrile
Vzorec: C₇H₄BrNO
MW: 198,017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527339
CAS číslo: 82380-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR