Tisk…

1-(Carboxymethyl)cyclohexane-1-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR908073-1G OR908073-250MG
APOSOR908073-1GEA 22950 CZK
APOSOR908073-1G APOSOR908073-250MG
1-(Carboxymethyl)cyclohexane-1-carboxylic acid 95%
1-(Carboxymethyl)cyclohexane-1-carboxylic acid
Vzorec: C₉H₁₄O₄
MW: 186,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037521
CAS číslo: 67950-95-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR