Tisk…

4-Formyl-3-methylbenzoic acid 97%

Dodavatel: Apollo Scientific

OR908061-1G OR908061-5G
APOSOR908061-1GEA 8270 CZK
APOSOR908061-1G APOSOR908061-5G
4-Formyl-3-methylbenzoic acid 97%
4-Formyl-3-methylbenzoic acid
Vzorec: C₉H₈O₃
MW: 164,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20660431
CAS číslo: 24078-23-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR