Tisk…

3-Chloro-2-ethylbenzoic acid 95%

Dodavatel: Apollo Scientific

OR908057-1G OR908057-250MG
APOSOR908057-1GEA 16340 CZK
APOSOR908057-1G APOSOR908057-250MG
3-Chloro-2-ethylbenzoic acid 95%
3-Chloro-2-ethylbenzoic acid
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18410965
CAS číslo: 67648-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR