Methyl 5-chloro-2-methylbenzoate 98%
Dodavatel: Apollo Scientific
OR908056-25G
OR908056-5G
APOSOR908056-25GEA
6520
CZK
APOSOR908056-25G
APOSOR908056-5G
Methyl 5-chloro-2-methylbenzoate 98%
Methyl 5-chloro-2-methylbenzoate
Vzorec:
C₉H₉ClO₂ MW: 184,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09831993 CAS číslo: 99585-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...