Tisk…

3-Cyano-N,N-dimethylbenzamide 97%

Dodavatel: Apollo Scientific

OR908045-1G OR908045-5G
APOSOR908045-1GEA 6220 CZK
APOSOR908045-1G APOSOR908045-5G
3-Cyano-N,N-dimethylbenzamide 97%
3-Cyano-N,N-dimethylbenzamide
Vzorec: C₁₀H₁₀N₂O
MW: 174,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07186286
CAS číslo: 33322-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR