Tisk…

2,4-Dioxo-4-phenylbutanoic acid 98%

Dodavatel: Apollo Scientific

OR908042-1G OR908042-5G
APOSOR908042-1GEA 4270 CZK
APOSOR908042-1G APOSOR908042-5G
2,4-Dioxo-4-phenylbutanoic acid 98%
2,4-Dioxo-4-phenylbutanoic acid
Vzorec: C₁₀H₈O₄
MW: 192,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00456054
CAS číslo: 5817-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR