Tisk…

4-Cyano-N,N-dimethylbenzamide 98%

Dodavatel: Apollo Scientific

OR908026-1G OR908026-250MG OR908026-5G
APOSOR908026-1GEA 11380 CZK
APOSOR908026-1G APOSOR908026-250MG APOSOR908026-5G
4-Cyano-N,N-dimethylbenzamide 98%
4-Cyano-N,N-dimethylbenzamide
Vzorec: C₁₀H₁₀N₂O
MW: 174,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07186293
CAS číslo: 24167-50-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR